Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-COX4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-39115SS NB110-39115
102121-838EA 208 USD InStock
102121-838 102121-826
Anti-COX4 Rabbit Polyclonal Antibody
Antibodies
The COX4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to COX4. This antibody reacts with human, mouse, rat, porcine, bovine, drosophila, insect, primate. The COX4 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: COX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Drosophila, Insect, Primate

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty