Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-ARNT/HIF-1 beta Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-110 NB100-110SS
102104-616EA 589.54 USD InStock
102104-616 102104-630
Anti-ARNT/HIF-1 beta Rabbit Polyclonal Antibody
Antibodies
The ARNT / HIF-1 beta Antibody from Novus Biologicals is a rabbit polyclonal antibody to ARNT / HIF-1 beta. This antibody reacts with human, mouse, rat, bovine, ferret, sheep. The ARNT / HIF-1 beta Antibody has been validated for the following applications: Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Supershift Assay.

Simple Western ,

Type: Primary
Antigen: ARNT/HIF-1 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Ferret, Sheep

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty