Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-NSP 5 alpha 3 alpha Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-518 NB100-518SS
102109-446EA 503.38 USD InStock
102109-446 102109-460
Anti-NSP 5 alpha 3 alpha Rabbit Polyclonal Antibody
Antibodies
The NSP 5 alpha 3 alpha Antibody from Novus Biologicals is a rabbit polyclonal antibody to NSP 5 alpha 3 alpha. This antibody reacts with human. The NSP 5 alpha 3 alpha Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: NSP 5 alpha 3 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty