Solid Gowning Benches, Electropolished Stainless Steel, Eagle MHC
Supplier: Eagle Group
CRB0936EP CRB0948EP CRB0960EP CRB0972EP CRB1236EP CRB1248EP CRB1260EP CRB1272EP CRB1284EP
58202-102EA 2571.33 USD InStock
58202-102 58202-604 58202-606 58202-608 58202-610 58202-612 58202-614 58202-616 58202-620
Solid Gowning Benches, Electropolished Stainless Steel, Eagle MHC
Furniture Gowning Benches
14 gauge Type 304 stainless steel top.
  • 17" overall height
  • Electropolished stainless steel finish
Front and rear feature a rolled edge with ends turned down at 90°. Flanged stainless foot plates for convenient attachment to floor. Shipped knock-down.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty