Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IFNB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
214-401-C40 214-401-C40S
RL214-401-C40EA 551.64 USD InStock
RL214-401-C40 RL214-401-C40S
Anti-IFNB1 Rabbit Polyclonal Antibody
Antibodies
This Protein A purified antibody is suitable for ELISA and Western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 19.5 kDa in size corresponding to swine IFN beta by western blotting.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:500-1:2000;

Type: Primary
Antigen: IFNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty