Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-POLB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-C65 600-401-C65S
RL600-401-C65EA 458.66 USD InStock
RL600-401-C65 RL600-401-C65S
Anti-POLB Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody is suitable for ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 38.1 kDa in size corresponding to POL-beta by western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:50000; Western Blot: 0.5 µg/ml;

Type: Primary
Antigen: POLB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty