Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexylphosphonic acid

Supplier: Origen Biomedical, Inc
Warning

044121-04 044121-09
AA44121-04EA 111 USD InStock
AA44121-04 AA44121-09
Hexylphosphonic acid
Hexylphosphonic acid
CAS No.: 4721-24-8 Molecular Formula: C6H13P(O)(OH)2 Formula Weight: 166.16 Physical Form: Powder Appearance: White to off-white Melting Point: 108-110° MDL No.: MFCD00013961


Formula: C₆H₁₅O₃P
MW: 166.16 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013961
CAS Number: 4721-24-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty