045525-06
045525-14
AA45525-06EA
111.66
USD
InStock
AA45525-06
AA45525-14
Indium(III) fluoride, anhydrous ≥99.95% (metals basis)
Indium(III) fluoride
Purity: 99.95%
CAS No.: 7783-52-0
Molecular Formula: F3In
Formula Weight: 171.82
Physical Form: Powder
Density: 4.39
Appearance: White
Odor: Odorless
Melting Point: ca 1170°
Boiling Point: 1200°
Merck Reference: 14,4959
MDL No.: MFCD00016149