Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium oxide superactivated neutral Grade I

Supplier: Origen Biomedical, Inc

045901-30 045901-A1
AA45901-30EA 52.1 USD InStock
AA45901-30 AA45901-A1
Aluminium oxide superactivated neutral Grade I
Aluminium oxide superactivated neutral
CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 50-200 Micron APS Powder Refractive Index: 1.765 Appearance: White Odor: Odorless Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424


Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 14,356
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty