Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cerium ≥99.9% (REO, rare earth oxide basis), powder < 200 mesh

Supplier: Origen Biomedical, Inc
Danger

045909-06 045909-14
AA45909-06EA 213 USD InStock
AA45909-06 AA45909-14
Cerium ≥99.9% (REO, rare earth oxide basis), powder < 200 mesh
Cerium
Purity: 99.9% CAS No.: 7440-45-1 Physical Form: Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Silver Odor: Odorless UN No.: 3078 MDL No.: MFCD00010924


Formula: Ce
MW: 140.12 g/mol
Boiling Pt: 3468
Melting Pt: 795
Density: 6.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010924
CAS Number: 7440-45-1
EINECS: 231-154-9
UN: 3078
ADR: 4.3 II
Order Now
Specification Test Results
Form Packaged under argon

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty