A10694-22
A10694-36
AAA10694-22EA
46.17
USD
InStock
AAA10694-22
AAA10694-36
Benzyl cyanide 98+%
Benzyl cyanide
Purity: 98+%
CAS No.: 140-29-4
Molecular Formula: C8H7N
Formula Weight: 117.15
Flash Point: 101°(213°F)
Physical Form: Liquid
Density: 1.015
Refractive Index: 1.5230
Appearance: Colorless
Melting Point: -24°
Boiling Point: 234°
Beilstein Reference: 385941
Merck Reference: 14,1131
UN No.: 2470
MDL No.: MFCD00001894