A11721-06
A11721-22
A11721-14
AAA11721-06EA
43.3
USD
InStock
AAA11721-06
AAA11721-22
AAA11721-14
Diphenylphosphinic chloride 97+%
Diphenylphosphinic chloride
Purity: 97+%
CAS No.: 1499-21-4
Molecular Formula: C12H10ClOP
Formula Weight: 236.64
Flash Point: 110°(230°F)
Refractive Index: 1.6090
Melting Point: ca 20°
Boiling Point: 210-212°/8mm
UN No.: 3265
MDL No.: MFCD00002077