Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Betaine hydrochloride 99%

Supplier: Origen Biomedical, Inc
Warning

A16122-0B A16122-30
AAA16122-0IEA 370.05 USD InStock
AAA16122-0I AAA16122-0B AAA16122-30
Betaine hydrochloride 99%
Betaine hydrochloride
An organic osmolyte

Purity: 99% CAS No.: 590-46-5 Molecular Formula: (CH3)3NCH2CO2.HCl Formula Weight: 153.61 Melting Point: ca 240° dec. Beilstein Reference: 3916181 Merck Reference: 14,1179 MDL No.: MFCD00011903


Formula: C₅H₁₂ClNO₂
MW: 153.61 g/mol
Melting Pt: ∼ 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011903
CAS Number: 590-46-5
EINECS: 209-683-1
Merck Index: 14,1179
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty