Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethoxy-5-methyl-p-benzoquinone 98%

Warning

B24777-06 B24777-03 B24777-14
AAAB24777-06EA 163.99 USD InStock
AAAB24777-06 AAAB24777-03 AAAB24777-14
2,3-Dimethoxy-5-methyl-p-benzoquinone 98%
2,3-Dimethoxy-5-methyl-p-benzoquinone
Purity: 98% CAS No.: 605-94-7 Molecular Formula: C9H10O4 Formula Weight: 182.18 Melting Point: 57-60° Beilstein Reference: 1640422


Formula: C₉H₁₀O₄
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00001595
CAS Number: 605-94-7
EINECS: 210-100-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty