Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Benzyloxyacetophenone 97%

Supplier: Origen Biomedical, Inc
Warning

B25084-06 B25084-14 B25084-22
AAB25084-06EA 45.6 USD InStock
AAB25084-06 AAB25084-14 AAB25084-22
3'-Benzyloxyacetophenone 97%
3'-Benzyloxyacetophenone
Purity: 97% CAS No.: 34068-01-4 Molecular Formula: C15H14O2 Formula Weight: 226.28 MDL No.: MFCD00026221


Formula: C₁₅H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00026221
CAS Number: 34068-01-4
EINECS: 251-816-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty