Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium hydrogen sulphate, anhydrous 90+% remainder mainly sodium sulphate

Supplier: Origen Biomedical, Inc
Danger

B25587-0B B25587-0I B25587-30
AAB25587-0BEA 82.22 USD InStock
AAB25587-0B AAB25587-0I AAB25587-30
Sodium hydrogen sulphate, anhydrous 90+% remainder mainly sodium sulphate
Sodium hydrogen sulphate
Purity: 90+% CAS No.: 7681-38-1 Molecular Formula: NaHSO4 Formula Weight: 120.06 Odor: Odorless pH: 1.0 Melting Point: 177-180° Merck Reference: 14,8586 UN No.: 3260 MDL No.: MFCD00003531


Formula: HNaO₄S
MW: 120.06 g/mol
Melting Pt: 181.85
Density: 2.435
Storage Temperature: Ambient
MDL Number: MFCD00003531
CAS Number: 7681-38-1
EINECS: 231-665-7
Merck Index: 14,8586
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty