Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Pentafluoroethyl)trimethylsilane 97%

Supplier: Origen Biomedical, Inc
Danger

H25775-03 H25775-06
AAH25775-03EA 257 USD InStock
AAH25775-03 AAH25775-06 AAH25775-14
(Pentafluoroethyl)trimethylsilane 97%
(Pentafluoroethyl)trimethylsilane


Formula: C₅H₉F₅Si
Storage Temperature: Ambient
MDL Number: MFCD04116436
CAS Number: 124898-13-1
UN: 1993
ADR: "3",III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty