H26247-03
H26247-06
H26247-MD
AAH26247-03EA
48.2
USD
InStock
AAH26247-03
AAH26247-06
AAH26247-MD
2-Bromobenzyl mercaptan 99%
2-Bromobenzyl mercaptan
Purity: 99%
CAS No.: 143888-85-1
Molecular Formula: C7H7BrS
Formula Weight: 203.10
Flash Point: 110°(230°F)
Refractive Index: 1.6170
Boiling Point: 256-257°
UN No.: 2810
MDL No.: MFCD04038925