H27140-06
H27140-14
AAH27140-06EA
55.6
USD
InStock
AAH27140-06
AAH27140-14
2,4-Dimethoxybenzylamine 98%
2,4-Dimethoxybenzylamine
Purity: 98%
CAS No.: 20781-20-8
Molecular Formula: C9H13NO2
Formula Weight: 167.21
Flash Point: 110°(230°F)
Refractive Index: 1.5490
Boiling Point: 140°/1mm
UN No.: 2735
MDL No.: MFCD00052393