Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromophenyl phenyl ether (BDE No. 3) 99%

Supplier: Origen Biomedical, Inc
Danger

L01473-14 L01473-22
AAAL01473-14EA 50.8 USD InStock
AAAL01473-14 AAAL01473-22
4-Bromophenyl phenyl ether (BDE No. 3) 99%
4-Bromophenyl phenyl ether (BDE No. 3)
Purity: 99% CAS No.: 101-55-3 Molecular Formula: C12H9BrO Formula Weight: 249.11 Flash Point: 110°(230°F) Refractive Index: 1.6080 Melting Point: 17-18° Boiling Point: 304-305° Beilstein Reference: 1308972 UN No.: 3082 MDL No.: MFCD00000094


Formula: C₁₂H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD00000094
CAS Number: 101-55-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty