L03239-09
L03239-18
AAAL03239-09EA
56.8
USD
InStock
AAAL03239-09
AAAL03239-18
Dichlorodiethylsilane 94%
Dichlorodiethylsilane
Purity: 94%
CAS No.: 1719-53-5
Molecular Formula: C4H10Cl2Si
Formula Weight: 157.12
Flash Point: 28°(82°F)
Refractive Index: 1.4310
Odor: Acrid
Melting Point: -97°
Boiling Point: 129-131°
Beilstein Reference: 605313
UN No.: 1767
MDL No.: MFCD00000493