Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allyltriethoxysilane 97%

Supplier: Origen Biomedical, Inc
Danger

L04297-14 L04297-06
AAAL04297-14EA 169 USD InStock
AAAL04297-14 AAAL04297-06
Allyltriethoxysilane 97%
Allyltriethoxysilane
Purity: 97% CAS No.: 2550-04-1 Molecular Formula: C9H20O3Si Formula Weight: 204.34 Flash Point: 21°(69°F) Refractive Index: 1.4070 Boiling Point: 176-177° Beilstein Reference: 1772306 UN No.: 1993 MDL No.: MFCD00009064


Formula: C₉H₂₀O₃Si
Boiling Pt: 176…177 °C
Density: 0.953
Flash Pt: 21 °C(69 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009064
CAS Number: 2550-04-1
EINECS: 219-843-2
UN: 1993
ADR: "3",III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty