Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chromotrope 2B ≥97%

Supplier: Origen Biomedical, Inc
Warning

L11330-14 L11330-22
AAAL11330-14EA 49.4 USD InStock
AAAL11330-14 AAAL11330-22
Chromotrope 2B ≥97%
Chromotrope 2B
CAS No.: 548-80-1 Molecular Formula: C16H9N3Na2O10S2 Formula Weight: 513.37 Melting Point: 300° Beilstein Reference: 4109733 Merck Reference: 14,2240 MDL No.: MFCD00003951


Formula: C₁₆H₉N₃Na₂O₁₀S₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003951
CAS Number: 548-80-1
EINECS: 208-959-9
Merck Index: 14,2240
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty