L13454-03
L13454-06
L13454-14
AAAL13454-03EA
32.7
USD
InStock
AAAL13454-03
AAAL13454-06
AAAL13454-14
4-Methyl-3-nitrobenzaldehyde 98%
4-Methyl-3-nitrobenzaldehyde
Purity: 98%
CAS No.: 31680-07-6
Molecular Formula: C8H7NO3
Formula Weight: 165.15
Flash Point: 110°(230°F)
Melting Point: 44-48°
Beilstein Reference: 1637706
MDL No.: MFCD00017011