Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Poly(methylhydrosiloxane)

Supplier: Origen Biomedical, Inc

L14561-30 L14561-0B L14561-18
AAAL14561-30EA 67.86 USD InStock
AAAL14561-30 AAAL14561-0B AAAL14561-18
Poly(methylhydrosiloxane)
Poly(methylhydrosiloxane)
Poly(methylhydrosiloxane) is used in the reduction of esters to alcohols, catalyzed by a combination of titanocene dichloride and either n-BuLi or EtMgBr. It is a safer alternative to triethoxysilane, B22063, for reduction of phosphine oxides to phosphines, catalyzed by Ti(O-i-Pr)4. It is also used in greener amine synthesis by reductive amination as an alternative to borohydrides.

CAS No.: 63148-57-2 Molecular Formula: (CH3)3SiO[(CH3)HSiO]nSi(CH3) Formula Weight: ca 1900 Flash Point: 204°(399°F) Refractive Index: 1.3980


Formula: (CH₃)₃SiO[(CH₃)HSiO]nSi(CH₃)
Boiling Pt: ? 140 °C (0.003 hPa)
Melting Pt: –55 °C
Density: 0.971 g/cm³ (25 °C)
Flash Pt: 204 °C(399 °F)
Storage Temperature: Ambient
MDL Number: MFCD00084478
CAS Number: 63148-57-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty