Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Iron sulfide ≥99.9%

Supplier: Origen Biomedical, Inc

012842-03 012842-09
AA12842-03EA 49.8 USD InStock
AA12842-03 AA12842-09 AA12842-18
Iron sulfide ≥99.9%
Iron sulfide
Purity: 99.9% CAS No.: 12068-85-8 Molecular Formula: FeS2 Formula Weight: 119.98 Physical Form: -100 Mesh Powder Density: 4.87 Odor: Odorless Melting Point: 1171° MDL No.: MFCD00064690


Formula: FeS₂
MW: 119.98 g/mol
Melting Pt: 1171 °C
Density: 4.87
Storage Temperature: Ambient
MDL Number: MFCD00064690
CAS Number: 12068-85-8
EINECS: 235-106-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty