Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc bromide hydrate ≥99.999% (metals basis), Puratronic®

Danger

012960-22 012960-06
AA12960-22EA 467.99 USD InStock
AA12960-22 AA12960-06
Zinc bromide hydrate ≥99.999% (metals basis), Puratronic®
Zinc bromide hydrate
Purity: 99.999% CAS No.: 7699-45-8 Molecular Formula: ZnBr2.xH2O (x≈2) Formula Weight: 225.19(anhy) Physical Form: Crystalline UN No.: 3260 MDL No.: MFCD00011294


Formula: Br₂Zn·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011294
CAS Number: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8,II
Merck Index: 14,10129
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty