Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc orthophosphate ≥99.995% (metals basis), Puratronic®

Supplier: Origen Biomedical, Inc

013013-14 013013-06
AA13013-14EA 322 USD InStock
AA13013-14 AA13013-06
Zinc orthophosphate ≥99.995% (metals basis), Puratronic®
Zinc orthophosphate
Purity: 99.995% CAS No.: 7779-90-0 Molecular Formula: O8P2Zn3 Formula Weight: 386.05 Physical Form: Powder Density: 3.75 Appearance: White Odor: Odorless UN No.: 3077 MDL No.: MFCD00036282


Formula: O₈P₂Zn₃
MW: 386.08 g/mol
Density: 3.75
Storage Temperature: Ambient
MDL Number: MFCD00036282
CAS Number: 7779-90-0
EINECS: 231-944-3
UN: 3077
ADR: 9 III
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty