Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc diethyldithiocarbamate (17 - 19,5% Zn)

Supplier: Origen Biomedical, Inc
Warning

018697-36 018697-A3
AA18697-36EA 35.7 USD InStock
AA18697-36 AAA18697-A3
Zinc diethyldithiocarbamate (17 - 19,5% Zn)
Zinc diethyldithiocarbamate
Applications: Chelating agent

CAS No.: 14324-55-1 Molecular Formula: C10H20N2S4Zn Formula Weight: 361.90 Flash Point: 204°(400°F) Physical Form: Crystalline Density: 1.48 Appearance: Off-white Odor: Odorless pH: 7.8 Melting Point: 178-183° UN No.: 3077 MDL No.: MFCD00064798

Applications: Chelating agent


Formula: C₅H₁₁NS₂
MW: 361.90 g/mol
Melting Pt: 178…183 °C
Density: 1.48
Flash Pt: 204 °C (400 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064798
CAS Number: 14324-55-1
EINECS: 238-270-9
UN: 3077
ADR: 9 III
Order Now
Specification Test Results
Form Crystalline

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty