Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Linear alkylbenzenesulfonic acid ≥97%

Supplier: Origen Biomedical, Inc
Danger

030574-A1 030574-D5 030574-30
AA30574-A1EA 94.2 USD InStock
AA30574-A1 AA30574-D5 AA30574-30
Linear alkylbenzenesulfonic acid ≥97%
Linear alkylbenzenesulfonic acid
Purity: 97% CAS No.: 68584-22-5 Molecular Formula: RC6H4SO3H Physical Form: Liquid UN No.: 2586 MDL No.: MFCD00147445


Formula: C₁₈H₃₀O₃S
Storage Temperature: Ambient
MDL Number: MFCD00147445
CAS Number: 68584-22-5
EINECS: 271-528-9
UN: 2586
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty