Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl azide ≥94%

Supplier: Origen Biomedical, Inc
Warning

032635-18 032635-04 032635-09
AA32635-18EA 702 USD InStock
AA32635-18 AA32635-04 AA32635-09
Benzyl azide ≥94%
Benzyl azide
Insoluble in water

Purity: 94% CAS No.: 622-79-7 Molecular Formula: C6H5CH2N3 Formula Weight: 133.15 Flash Point: 43°(110°F) Physical Form: Liquid Density: 1.065 Refractive Index: 1.5341 Appearance: Colorless to pale brown Boiling Point: 82-85°/16mm Solubility: Insoluble in water UN No.: 1993 MDL No.: MFCD00013836


Formula: C₇H₇N₃
MW: 133.154 g/mol
Boiling Pt: 82…85 °C (16 mmHg)
Density: 1.065
Flash Pt: 43 °C (110 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013836
CAS Number: 622-79-7
UN: 1993
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty