039329-14
039329-06
AA39329-14EA
227
USD
InStock
AA39329-14
AA39329-06
Lithium cyclopentadienide ≥97%
Lithium cyclopentadienide
Purity: 97%
CAS No.: 16733-97-4
Molecular Formula: C5H5Li
Formula Weight: 72.04
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
UN No.: 3180
MDL No.: MFCD00001439