Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl arachidate 98+%

Supplier: Origen Biomedical, Inc

042399-09 042399-04
AA42399-09EA 271 USD InStock
AA42399-09 AA42399-04
Methyl arachidate 98+%
Methyl arachidate
Purity: 98+% CAS No.: 1120-28-1 Molecular Formula: CH3(CH2)18COOCH3 Formula Weight: 326.56 Flash Point: 110°(230°F) Physical Form: Crystalline powder Melting Point: 48° Boiling Point: 215°/10mm MDL No.: MFCD00009014


Formula: C₂₁H₄₂O₂
MW: 326.56 g/mol
Boiling Pt: 215 °C (10 mmHg)
Melting Pt: 48 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009014
CAS Number: 1120-28-1
EINECS: 214-304-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty