Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-cyanoacetoacetate

Supplier: Origen Biomedical, Inc
Danger

043792-09 043792-04
AA43792-09EA 157 USD InStock
AA43792-09 AA43792-04
Ethyl-2-cyanoacetoacetate
Ethyl-2-cyanoacetoacetate
Product exists as a 1:2 keto:enol tautomer mixture

CAS No.: 634-55-9 Molecular Formula: C2H5OOCCH(CN)COCH3 Formula Weight: 155.16 Physical Form: Liquid UN No.: 3276 MDL No.: MFCD00043487


Formula: C₇H₉NO₃
MW: 155.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043487
CAS Number: 634-55-9
UN: 3276
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty