Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hafnium(IV) sulfate ≥99.9% (max. 1% Zr)

Supplier: Origen Biomedical, Inc

044244-14 044244-06
AA44244-14EA 1171 USD InStock
AA44244-14 AA44244-06
Hafnium(IV) sulfate ≥99.9% (max. 1% Zr)
Hafnium(IV) sulfate
Purity: 99.9% CAS No.: 15823-43-5 Molecular Formula: Hf(SO4)2 Formula Weight: 370.62(anhy) Physical Form: Powder MDL No.: MFCD00168081


Formula: Hf(SO₄)₂
MW: 370.62 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00168081
CAS Number: 15823-43-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty