Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zeolite β, ammonium

Supplier: Origen Biomedical, Inc

045874-14 045874-22 045874-36
AA45874-14EA 29.3 USD InStock
AA45874-14 AA45874-22 AA45874-36
Zeolite β, ammonium
Zeolite beta, ammonium
Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)

CAS No.: 1318-02-1 Molecular Formula: SiO2:Al2O3 Physical Form: Powder, S.A. 710 m2/g, 38:1 mole ratio Appearance: White MDL No.: MFCD00132601

Applications: Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)


Formula: SiO₂:Al₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00132601
CAS Number: 1318-02-1
EINECS: 215-283-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty