Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium cyclopentadienide 2-3 M in THF

Supplier: Origen Biomedical, Inc
Danger

088300-JQ 088300-JT
AA88300-JQEA 90.9 USD InStock
AA88300-JQ AA88300-JT
Sodium cyclopentadienide 2-3 M in THF
Sodium cyclopentadienide
Reagent for the preparation of metallocenes

CAS No.: 4984-82-1 Molecular Formula: C5H5Na Formula Weight: 88.09 Flash Point: -17°(1°F) Physical Form: Liquid Density: 0.938 Sensitivity: Air and Moisture Sensitive Appearance: Pink to red Odor: Ether-like UN No.: 3399 MDL No.: MFCD00012161

Applications: Reagent for the preparation of metallocenes


Formula: C₅H₅Na
MW: 88.085 g/mol
Density: 0.938
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012161
CAS Number: 4984-82-1
EINECS: 225-636-8
UN: 3399
ADR: 4.3 II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty