Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-m-toluic acid 98%

Supplier: Origen Biomedical, Inc
Warning

A12280-03 A12280-06
AAA12280-03EA 23.4 USD InStock
AAA12280-03 AAA12280-06
2-Fluoro-m-toluic acid 98%
2-Fluoro-m-toluic acid
Purity: 98% CAS No.: 315-31-1 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Melting Point: 114-116° MDL No.: MFCD00042504


Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00042504
CAS Number: 315-31-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty