Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Phosphonooxy)benzoic acid 98%

Supplier: Origen Biomedical, Inc
Warning

A14417-06 A14417-03
AAA14417-06EA 379 USD InStock
AAA14417-06 AAA14417-14 AAAA14417-03
2-(Phosphonooxy)benzoic acid 98%
2-(Phosphonooxy)benzoic acid
Purity: 98% CAS No.: 6064-83-1 Molecular Formula: C7H7O6P Formula Weight: 218.10 Melting Point: 162-166° Beilstein Reference: 2619142 MDL No.: MFCD00042645


Formula: C₇H₇O₆P
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00042645
CAS Number: 6064-83-1
EINECS: 227-993-5
Merck Index: 14,4255
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty