A16006-06
A16006-14
A16006-03
AAA16006-06EA
211
USD
InStock
AAA16006-06
AAA16006-14
AAA16006-03
cis-Aconitic anhydride 98%
cis-Aconitic anhydride
Purity: 98%
CAS No.: 6318-55-4
Molecular Formula: C6H4O5
Formula Weight: 156.10
Storage Temperature: 2°C to 8°C
Melting Point: 74-78°
Beilstein Reference: 127575
UN No.: 3261
MDL No.: MFCD00005524