Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diiodofluorescein

Supplier: Origen Biomedical, Inc

A16554-18 A16554-09
AAA16554-18EA 111 USD InStock
AAA16554-18 AAA16554-09
Diiodofluorescein
Diiodofluorescein
CAS No.: 31395-16-1 Molecular Formula: C20H10I2O5 Formula Weight: 584.11 Melting Point: 239-241° dec. MDL No.: MFCD00036461


Formula: C₂₀H₁₀I₂O₅
Melting Pt: 239…241 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00036461
CAS Number: 31395-16-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty