Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrophenylacetone 98%

Supplier: Origen Biomedical, Inc
Warning

B21183-06 B21183-14 B21183-03
AAB21183-06EA 180 USD InStock
AAB21183-06 AAB21183-14 AAB21183-03
4-Nitrophenylacetone 98%
4-Nitrophenylacetone
Purity: 98% CAS No.: 5332-96-7 Molecular Formula: C9H9NO3 Formula Weight: 179.18 Melting Point: 61-64° Beilstein Reference: 1953054


Formula: C₉H₉NO₃
Melting Pt: 61…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00051518
CAS Number: 5332-96-7
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty