Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5,6-Triaminopyrimidine sulfate hydrate, (max. 8% H₂O) ≥98%

Supplier: Origen Biomedical, Inc
Warning

B21369-03 B21369-06
AAB21369-03EA 50.3 USD InStock
AAB21369-03 AAB21369-06
4,5,6-Triaminopyrimidine sulfate hydrate, (max. 8% H₂O) ≥98%
4,5,6-Triaminopyrimidine sulfate hydrate
Purity: 98+% CAS No.: 207742-76-5 Molecular Formula: C4H7N5.H2SO4.xH2O Formula Weight: 223.22(anhy) Melting Point: 300° MDL No.: MFCD00012789


Formula: C₄H₇N₅·H₂SO₄·xH₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012789
CAS Number: 207742-76-5
EINECS: 256-446-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty