Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethylhydrocupreine hydrochloride 97%

Supplier: Origen Biomedical, Inc

B21627-ME B21627-MC
AAB21627-MEEA 108 USD InStock
AAB21627-ME AAB21627-MC
Ethylhydrocupreine hydrochloride 97%
Ethylhydrocupreine hydrochloride
Purity: 97% CAS No.: 3413-58-9 Molecular Formula: C21H26N2O2.HCl Formula Weight: 374.91 Storage Temperature: 2°C to 8°C Melting Point: ca 240° dec. MDL No.: MFCD00135594


Formula: C₂₁H₂₆N₂O₂·HCl
MW: 374.91 g/mol
Melting Pt: ∼ 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00135594
CAS Number: 3413-58-9
EINECS: 222-302-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty