Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(1,3-Thiazol-2-yl)propan-1-one 99%

Supplier: Origen Biomedical, Inc

B21711-04 B21711-09
AAB21711-04EA 57.5 USD InStock
AAB21711-04 AAB21711-09
1-(1,3-Thiazol-2-yl)propan-1-one 99%
1-(1,3-Thiazol-2-yl)propan-1-one
Purity: 99% CAS No.: 43039-98-1 Molecular Formula: C6H7NOS Formula Weight: 141.19 Flash Point: 100°(212°F) Boiling Point: 110°/5mm MDL No.: MFCD01681404


Formula: C₆H₇NOS
Boiling Pt: 110 °C/5 mm
Density: 1.174
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD01681404
CAS Number: 43039-98-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty