Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium iron(III) hexacyanoferrate(II) hydrate, tech.


B21908-14 B21908-22
AAB21908-14EA 55.19 USD InStock
AAB21908-14 AAB21908-22
Ammonium iron(III) hexacyanoferrate(II) hydrate, tech.
Ammonium iron(III) hexacyanoferrate(II) hydrate
CAS No.: 25869-00-5 Molecular Formula: (NH4)Fe[Fe(CN)6].xH2O Formula Weight: 285.83(anhy) MDL No.: MFCD01726733


Formula: (NH₄)Fe[Fe(CN)₆]·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD01726733
CAS Number: 25869-00-5
EINECS: 247-304-1
Merck Index: 14,7910
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty