B22144-06
B22144-14
AAB22144-06EA
38.3
USD
InStock
AAB22144-06
AAB22144-14
S-Methyl thiobutyrate 98%
S-Methyl thiobutyrate
Purity: 98%
CAS No.: 2432-51-1
Molecular Formula: C5H10OS
Formula Weight: 118.20
Flash Point: 34°(93°F)
Refractive Index: 1.4620
Boiling Point: 142-143°
Beilstein Reference: 1848987
UN No.: 1993
MDL No.: MFCD00009872