B22292-06
B22292-14
AAB22292-06EA
68.69
USD
InStock
AAB22292-06
AAB22292-14
Perfluorodecanoic acid 97%
Perfluorodecanoic acid
Purity: 97%
CAS No.: 335-76-2
Molecular Formula: C10HF19O2
Formula Weight: 514.09
Melting Point: 77-82°
Boiling Point: 218°
UN No.: 2923
MDL No.: MFCD00004175