B22616-06
B22616-14
AAB22616-06EA
79.5
USD
InStock
AAB22616-06
AAB22616-14
4-Methyloctanoic acid 98%
4-Methylcaprylic acid
Purity: 98%
CAS No.: 54947-74-9
Molecular Formula: C9H18O2
Formula Weight: 158.24
Flash Point: 110°(230°F)
Refractive Index: 1.4335
Boiling Point: 272°
Beilstein Reference: 1755426
UN No.: 3265
MDL No.: MFCD00051938