Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Julolidine 98%

Supplier: Origen Biomedical, Inc

B22976-03 B22976-14 B22976-06
AAB22976-03EA 32.1 USD InStock
AAB22976-03 AAB22976-14 AAB22976-06
Julolidine 98%
Julolidine
Purity: 98% CAS No.: 479-59-4 Molecular Formula: C12H15N Formula Weight: 173.26 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 32-37° Boiling Point: 170-173°/31mm Beilstein Reference: 139925 MDL No.: MFCD00006917


Formula: C₁₂H₁₅N
MW: 173.26 g/mol
Boiling Pt: 170…173 °C/31 mm
Melting Pt: 32…37 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006917
CAS Number: 479-59-4
EINECS: 207-535-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty